missing translation for 'onlineSavingsMsg'
Learn More

cis-8,11,14-Eicosatrienoic Acid, TRC

Product Code. 30009105
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009105 100 mg 100mg
17977154 10 mg 10mg
17988314 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009105 Supplier Toronto Research Chemicals Supplier No. TRCE477930100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1783-84-2
Molecular Formula C20 H34 O2
Molecular Weight (g/mol) 306.48
Synonym 8,11,14-Eicosatrienoic acid, (Z,Z,Z)- (8CI), (8Z,11Z,14Z)-8,11,14-Eicosatrienoic acid, (Z,Z,Z)-8,11,14-Eicosatrienoic acid, 8,11,14-Eicosatrienoic acid, 8,11,14-all-cis-Eicosatrienoic acid, 8Z,11Z,14Z-Eicosatrienoic acid, Bishomo-γ-linolenic acid, DGLA, Dihomo-γ-linolenic acid, Diroleuton, Ro 12-1989, all-cis-8,11,14-Eicosatrienoic acid, cis-8,11,14-Eicosatrienoic acid, cis-8,cis-11,cis-14-Eicosatrienoic acid, γ-Homolinolenic acid
IUPAC Name (8Z,11Z,14Z)-icosa-8,11,14-trienoic acid
SMILES CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13H,2-5,8,11,14-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-
Formula Weight 306.2559
Chemical Name or Material all-cis-8,11,14-Eicosatrienoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.