missing translation for 'onlineSavingsMsg'
Learn More

cis-6-(Benzyloxycarbonylamino)-3-cyclohexene-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15437686
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437686 250 mg 250mg
15427686 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437686 Supplier Thermo Scientific Alfa Aesar Supplier No. H62650.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124753-65-7
Molecular Formula C15H16NO4
Molecular Weight (g/mol) 274.30
MDL Number MFCD01863246,MFCD01863246,MFCD03844603
InChI Key QIAAZWPFGHTXNZ-OLZOCXBDSA-M
Synonym 1s,6r-6-benzyloxy carbonyl amino cyclohex-3-ene-1-carboxylic acid, cis-6-benzyloxycarbonylaminocyclohex-3-enecarboxylic acid, cis-6-benzyloxy carbonyl amino cyclohex-3-enecarboxylic acid, 1s-6beta-benzyloxycarbonylamino-3-cyclohexene-1beta-carboxylic acid, 3-cyclohexene-1-carboxylic acid, 6-phenylmethoxy carbonyl amino-, 1r,6s-rel-9ci
PubChem CID 21356434
IUPAC Name (1S,6R)-6-(phenylmethoxycarbonylamino)cyclohex-3-ene-1-carboxylic acid
SMILES [O-]C(=O)[C@@H]1CC=CC[C@@H]1NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 275.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material cis-6-(Benzyloxycarbonylamino)-3-cyclohexene-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.