missing translation for 'onlineSavingsMsg'
Learn More

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%

Product Code. 15400656
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15400656

Brand: Thermo Scientific Alfa Aesar H53446.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2746-19-2 | C9H8O3 | 164.16 g/mol

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2746-19-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00630580
InChI Key KNDQHSIWLOJIGP-RNGGSSJXSA-N
Synonym cis-5-norbornene-exo-2,3-dicarboxylic anhydride, 3ar,4r,7s,7as-rel-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, himic anhydride, unii-n1kqo6559b, norborn-5-ene-2exo,3exo-dicarboxylic acid anhydride, 4,7-methanoisobenzofuran-1,3-dione, 3a,4,7,7a-tetrahydro-, 3aalpha,4beta,7beta,7aalpha-9ci, exo-5-norbornene-2,3-dicarboxylic anhydride, exo-3,6-methylene-1,2,3,6-tetrahydrophthalic anhydride, endicanhydride, 1r,2r,6s,7s-4-oxatricyclo 5.2.1.0 2 ,? dec-8-ene-3,5-dione
PubChem CID 637794
SMILES C1C2C=CC1C3C2C(=O)OC3=O

Specifications

Melting Point 140°C to 145°C
Quantity 25 g
Formula Weight 164.16
Percent Purity 95%
Chemical Name or Material cis-5-Norbornene-exo-2,3-dicarboxylic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.