missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-L-proline, TRC

Product Code. 30401508
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401508 100 mg
30389328 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401508 Supplier Toronto Research Chemicals Supplier No. TRCH952375100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-27-9
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
Synonym (4S)-4-Hydroxy-L-proline, (2S,4S)-4-Hydroxyproline, (S)-Allo-hydroxyproline, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic Acid, L-Allohydroxyproline, cis-4-Hydroxyproline, cis-Hydroxyproline
IUPAC Name (2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@@H]1CN[C@H](C(O)=O)C1

Specifications

InChI Formula InChI=1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4-/m0/s1
Quantity 100 mg
Formula Weight 131.0582
Shipping Condition Room Temperature
Chemical Name or Material cis-4-Hydroxy-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.