missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-L-proline methyl ester hydrochloride, 95%

Product Code. p-7041509
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15417366

Brand: Thermo Scientific Alfa Aesar H62384.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40126-30-5 | C6H12ClNO3 | 181.616 g/mol

cis-4-Hydroxy-L-proline methyl ester hydrochloride is an Intermediate in pharmaceutical and organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-Hydroxy-L-proline methyl ester hydrochloride is an Intermediate in pharmaceutical and organic syntheses.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40126-30-5
Molecular Formula C6H12ClNO3
Molecular Weight (g/mol) 181.616
MDL Number MFCD00237835
InChI Key KLGSHNXEUZOKHH-FHAQVOQBSA-N
Synonym methyl 2s,4s-4-hydroxypyrrolidine-2-carboxylate hydrochloride, cis-4-hydroxy-l-proline methyl ester hydrochloride, 2s,4s-methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, 2s,4s-methyl 4-hydroxypyrrolidine-2-carboxylate-hcl, 2s,4s-methyl 4-hydroxypyrrolidine-2-carboxylate hcl, l-proline, 4-hydroxy-, methyl ester, hydrochloride, 4s, h-cis-hyp-ome hcl, h-cis-hyp-ome . hcl, cis-4-hydroxyproline methyl ester hcl salt, methylcis-4-hydroxy-l-prolinehydrochloride
PubChem CID 12764092
IUPAC Name methyl (2S,4S)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
SMILES COC(=O)C1CC(CN1)O.Cl

Specifications

Quantity 250 mg
Formula Weight 181.62
Percent Purity 95%
Chemical Name or Material cis-4-Hydroxy-L-proline methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.