missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-L-proline-d3 (Major), TRC

Product Code. 30400894
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400894 100 mg 100mg
30389993 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400894 Supplier Toronto Research Chemicals Supplier No. TRCH952379100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C5 D3 H6 N O3
Molecular Weight (g/mol) 134.148
Synonym cis-4-Hydroxy-L-proline-2,5,5-d3, (4S)-4-Hydroxy-L-proline-2,5,5-D3, cis-4-Hydroxy-L-proline-2,5,5-D3, (2S,4S)-4-Hydroxyproline-2,5,5-d3, (2S,4S)-4-Hydroxypyrrolidine-2,5,5-d3 acid
IUPAC Name (2S,4S)-2,5,5-trideuterio-4-hydroxypyrrolidine-2-carboxylic acid
SMILES [2H]C1([2H])N[C@@]([2H])(C[C@@H]1O)C(=O)O

Specifications

InChI Formula InChI=1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4-/m0/s1/i2D2,4D
Quantity 100 mg
Formula Weight 134.077
Shipping Condition Room Temperature
Chemical Name or Material cis-4-Hydroxy-L-proline-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.