missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Heptenal, 96%

Product Code. 10247560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10247560 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10247560 Supplier Thermo Scientific Acros Supplier No. 298220100
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6728-31-0 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6728-31-0
Color Colorless to Yellow
Density 0.8490g/mL
Boiling Point 46.0°C (13.0 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12O
Linear Formula CH3CH2CH=CHCH2CH2CHO
Refractive Index 1.4320 to 1.4360
MDL Number MFCD00014682
Quantity 10 mL
Specific Gravity 0.849
Synonym cis-4-heptenal, cis-4-hepten-1-al, z-hept-4-enal, z-4-heptenal, 4-heptenal, z, z-4-hepten-1-al, hept-cis-4-enal, unii-5bj99wwp64, 4-z-heptenal, hept-4 z-enal
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,propylene glycol
InChI Key VVGOCOMZRGWHPI-ARJAWSKDSA-N
SMILES CCC=CCCC=O
IUPAC Name (Z)-hept-4-enal
Molecular Weight (g/mol) 112.17
PubChem CID 5362814
Formula Weight 112.17
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material cis-4-Heptenal, 96%
Show More Show Less
compliance-icons
Product Identifier
  • cis-4-Heptenal
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.