missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%

Product Code. 11337406
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11337406

Brand: Thermo Scientific Alfa Aesar L09392.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2305-26-2 | C8H10O4 | 170.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2305-26-2
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00070482
InChI Key ILUAAIDVFMVTAU-OLQVQODUSA-N
Synonym cis-4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 4-cyclohexene-1,2-dicarboxylic acid, cis, cis-delta4-tetrahydrophthalic acid, 4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel, rel-1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-rel-4-cyclohexene-1,2-dicarboxylic acid, cis-.delta.4-tetrahydrophthalic acid, cis-.delta.4-cyclohexene-1,2-dicarboxylic acid
PubChem CID 504243
IUPAC Name (1S,2R)-cyclohex-4-ene-1,2-dicarboxylic acid
SMILES C1C=CCC(C1C(=O)O)C(=O)O

Specifications

Melting Point 165°C to 170°C
Quantity 100 g
Beilstein 2050419
Formula Weight 170.17
Percent Purity 98%
Chemical Name or Material cis-4-Cyclohexene-1,2-dicarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.