missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4-Aminocyclohexanol Hydrochloride, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | cis-4-Aminocyclohexanol Hydrochloride |
| CAS | 56239-26-0 |
| Molecular Formula | C6 H13 N O . Cl H |
| Quantity | 10 g |
| Synonym | cis-4-Aminocyclohexanol Hydrochloride, cis-4-Hydroxycyclohexylamine Hydrochloride |
| InChI Formula | InChI=1S/C6H13NO.ClH/c7-5-1-3-6(8)4-2-5;/h5-6,8H,1-4,7H2;1H/t5-,6+; |
| SMILES | Cl.N[C@@H]1CC[C@H](O)CC1 |
| IUPAC Name | 4-aminocyclohexan-1-ol;hydrochloride |
| Molecular Weight (g/mol) | 151.63 |
| Formula Weight | 151.0764 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction