missing translation for 'onlineSavingsMsg'
Learn More

cis-4,7,10,13,16,19-Docosahexaenoic Acid Methyl Ester, TRC

Product Code. 30382247
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382247 10 mg
30385866 100 mg
30377991 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382247 Supplier Toronto Research Chemicals Supplier No. TRCD49449510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2566-90-7
Molecular Formula C23 H34 O2
Molecular Weight (g/mol) 342.51
Synonym 4,7,10,13,16,19-Docosahexaenoic acid, methyl ester, (all-Z)- (8CI), Docosahexaenoic acid methyl ester, Methyl 4,7,10,13,16,19-cis-docosahexanenoate, Methyl all-cis-4,7,10,13,16,19-docosahexaenoate, Methyl cis,cis,cis,cis,cis,cis-docosa-4,7,10,13,16,19-hexaenoate, Methyl cis-4,7,10,13,16,19-docosahexaenoate, Methyl docosahexaenoate, all cis-DHA methyl ester, cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester
IUPAC Name methyl (4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoate
SMILES CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(=O)OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h4-5,7-8,10-11,13-14,16-17,19-20H,3,6,9,12,15,18,21-22H2,1-2H3/b5-4-,8-7-,11-10-,14-13-,17-16-,20-19-
Formula Weight 342.2559
Percent Purity >95%
Chemical Name or Material Docosahexaenoic Acid Methyl Ester (DHA ME)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.