missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%

Product Code. 11904971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11904971 1 g Glass bottle 1g
11914971 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11904971 Supplier Thermo Scientific Chemicals Supplier No. 427640010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6217-54-5 | C22H32O2 | 328.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.49
InChI Key MBMBGCFOFBJSGT-OBOJEMQYSA-N
PubChem CID 57417355
IUPAC Name (4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O

Specifications

Melting Point -44°C
Color Colorless to Yellow
Density 0.9500g/mL
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2CHCH)6CH2CH2COOH
Quantity 1 g
Merck Index 15, 3443
Specific Gravity 0.95
Synonym cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
Formula Weight 328.49
Percent Purity 98%
Physical Form Oil
Chemical Name or Material cis-4,7,10,13,16,19-Docosahexaenoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.