missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Octen-1-ol, 95%

Product Code. 11499316
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11499316

Brand: Thermo Scientific Alfa Aesar A19014.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20125-84-2 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20125-84-2
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014062
InChI Key YDXQPTHHAPCTPP-WAYWQWQTSA-N
Synonym cis-3-octen-1-ol, z-3-octen-1-ol, z-oct-3-en-1-ol, 3-octen-1-ol, z, unii-34a1x2y8m9, 3z-octen-1-ol, 3z-3-octen-1-ol, 3-octenol, cis, fema no. 3467, 3-octen-1-ol, 3z
PubChem CID 5364519
IUPAC Name (Z)-oct-3-en-1-ol
SMILES CCCCC=CCCO

Specifications

Density 0.85
Boiling Point 88°C to 89°C (15 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.447
Quantity 10 g
Beilstein 1720075
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material cis-3-Octen-1-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.