missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals

Product Code. 11429796
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429796 5 g 5g
11439796 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429796 Supplier Thermo Scientific Alfa Aesar Supplier No. B20288.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24965-90-0 | Molecular Formula: C7H14O | Molecular Weight: 114.188 g/mol

cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.

Solubility
Soluble in ethanol, ether. Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24965-90-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00067155
InChI Key HTSABYAWKQAHBT-RQJHMYQMSA-N
Synonym cis-3-methylcyclohexanol, cyclohexanol, 3-methyl-, cis, 1s,3r-3-methylcyclohexan-1-ol, 3-methyl cyclohexanol, cis, 3-methylcyclohexanol, z, cis 3-methylcyclohexanol, cis-3-methyl-cyclohexanol, 1s,3r-3-methyl-cyclohexanol, 1alpha-3alpha-methylcyclohexanol
PubChem CID 229503
IUPAC Name (1S,3R)-3-methylcyclohexan-1-ol
SMILES CC1CCCC(C1)O

Specifications

Flash Point 73°C (163°F)
Quantity 5 g
Solubility Information Soluble in ethanol,ether. Slightly soluble in water.
Formula Weight 114.19
Percent Purity 95%
Chemical Name or Material cis-3-Methylcyclohexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.