missing translation for 'onlineSavingsMsg'
Learn More

cis-3'-Hydroxy Cotinine, TRC

Product Code. 30398873
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398873 10 mg -
30398353 5 mg -
30405585 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398873 Supplier Toronto Research Chemicals Supplier No. TRCH92450510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37096-14-3
Molecular Formula C10 H12 N2 O2
Molecular Weight (g/mol) 192.21
Synonym 2-Pyrrolidinone, 3-hydroxy-1-methyl-5-(3-pyridinyl)-, (3S,5S)-, 2-Pyrrolidinone, 3-hydroxy-1-methyl-5-(3-pyridinyl)-, (3S-cis)-, (3S-cis)-3-Hydroxycotinine, cis-3'-Hydroxycotinine
IUPAC Name (3S,5S)-3-hydroxy-1-methyl-5-pyridin-3-ylpyrrolidin-2-one
SMILES CN1[C@@H](C[C@H](O)C1=O)c2cccnc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H12N2O2/c1-12-8(5-9(13)10(12)14)7-3-2-4-11-6-7/h2-4,6,8-9,13H,5H2,1H3/t8-,9-/m0/s1
Formula Weight 192.0899
Chemical Name or Material (3S,5S)-3'-Hydroxycotinine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.