missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(Benzyloxymethyl)cyclobutanol, 97%, Thermo Scientific Chemicals

Product Code. p-7041560
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409098 250 mg 250mg
15499088 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409098 Supplier Thermo Scientific Alfa Aesar Supplier No. H62579.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

cis-3-(Benzyloxymethyl)cyclobutanol is generally used as a intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-(Benzyloxymethyl)cyclobutanol is generally used as a intermediate in chemical research.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172324-68-4
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD06657676
InChI Key KVQOEODTOJCIPA-UHFFFAOYSA-N
Synonym cis-3-benzyloxymethylcyclobutanol, trans-3-benzyloxymethylcyclobutanol, trans-3-benzyloxy methyl cyclobutanol, cis-3-benzyloxy methyl cyclobutanol, 1r,3r-3-benzyloxy methyl cyclobutanol, cyclobutanol,3-phenylmethoxy methyl-, cis, 3-benzyloxy methyl cyclobutan-1-ol, 1r,3r-3-benzyloxy methyl cyclobutan-1-ol
PubChem CID 10442570
IUPAC Name 3-(phenylmethoxymethyl)cyclobutan-1-ol
SMILES C1C(CC1O)COCC2=CC=CC=C2

Specifications

Quantity 250 mg
Formula Weight 192.25
Percent Purity 97%
Chemical Name or Material cis-3-(Benzyloxymethyl)cyclobutanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.