missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-cyclopropanecarboxylic Acid, TRC

Product Code. 30134892
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134892 10 g 10g
30007014 1 g 1g
17972885 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134892 Supplier Toronto Research Chemicals Supplier No. TRCC42200010G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72748-35-7
Molecular Formula C9 H10 Cl F3 O2
Molecular Weight (g/mol) 242.62
Synonym Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, (1R,3R)-rel- (9CI), Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, cis-, rel-(1R,3R)-3-(2-Chloro-3,3,3-trifluoro-1-propen-1-yl)-2,2-dimethylcyclopropanecarboxylic acid
IUPAC Name (1R,3R)-3-(2-chloro-3,3,3-trifluoroprop-1-enyl)-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(C)[C@@H](C=C(Cl)C(F)(F)F)[C@H]1C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H10ClF3O2/c1-8(2)4(6(8)7(14)15)3-5(10)9(11,12)13/h3-4,6H,1-2H3,(H,14,15)/t4-,6-/m0/s1
Formula Weight 242.0321
Chemical Name or Material Tefluthrin (free acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.