missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ cis-22a

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
cis-22a is a TRPV6 inhibitor (IC50 = 0.32 μM), which exhibits selectivity against related TRPV channels and calcium channels. cis-22a displays antiproliferative effects on T47D human breast cancer cells.
Specifications
Specifications
| Chemical Name or Material | 1-[cis-4-(3-Methylphenyl)cyclohexyl]-4-(3-pyridinyl)piperazine ditrifluoroacetate |
| CAS | 1819366-84-1 |
| Quantity | 50 mg |
| Target | TRPV Inhibitors |
| Molecular Formula | C22H29N30.2 CF3CO2H |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction