missing translation for 'onlineSavingsMsg'
Learn More

cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%

Product Code. 10491772 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10491772 1 g Glass Bottle 1g
10197553 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10491772 Supplier Thermo Scientific Acros Supplier No. 334270010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63216-49-9 | C12H20NO4 | 242.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63216-49-9
Molecular Formula C12H20NO4
Molecular Weight (g/mol) 242.30
MDL Number MFCD01863244,MFCD06410968
InChI Key QJEQJDJFJWWURK-BDAKNGLRSA-M
Synonym 1r,2s-boc-achc, boc-1,2-cis-achc-oh, boc-cis-2-aminocyclohexanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2s-boc-2-aminocyclohexane carboxylic acid, n-boc-+/--cis-2-amino-cyclohexane-carboxylic acid, cis-2-tert-butoxycarbonylamino-cyclohexanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, cis-1-t-butyloxycarbonylamino-cyclohexyl-2-carboxylic acid, 1r,2s-2-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid
PubChem CID 1512487
IUPAC Name (1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)N[C@H]1CCCC[C@H]1C([O-])=O

Specifications

Melting Point 127°C to 131°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOOCC6H10NHCOOC(CH3)3
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform
Quantity 1 g
Flash Point 194°C
Formula Weight 243.3
Percent Purity 98%
Physical Form Amorphous Powder
Chemical Name or Material cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • cis-2-(tert-Butoxycarbonylamino)-cyclohexanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.