missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Specifications
Melting Point | 90.0°C to 94.0°C |
Color | White |
Assay Percent Range | 98% |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Formula Weight | 229.28 |
Percent Purity | 98% |
Quantity | 1 g |
Physical Form | Fine Crystalline Powder |
Chemical Name or Material | cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'