missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Pentene, Thermo Scientific Chemicals

Catalog Number 10320684
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
627-20-3
C5H10
70.14
MFCD00063978
QMMOXUPEWRXHJS-HYXAFXHYSA-N
cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z
5326160
(Z)-pent-2-ene
CC\C=C/C
This item is not returnable. View return policy
627-20-3
C5H10
70.14
MFCD00063978
QMMOXUPEWRXHJS-HYXAFXHYSA-N
cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z
5326160
(Z)-pent-2-ene
CC\C=C/C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -151.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 37.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3CHCHCH2CH3
Refractive Index 1.3810 to 1.3830 (20°C, 589nm)
Quantity 1 g
Merck Index 15, 7235
Specific Gravity 0.65
Solubility Information Solubility in water: insoluble.
Formula Weight 70.13
Percent Purity ≥95.0%
Physical Form Liquid
Chemical Name or Material cis-2-Pentene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • cis-2-Pentene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'