Learn More
cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer
Description
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Spécification
Spécification
| Density | 0.853 |
| Boiling Point | 138°C |
| Flash Point | 48°C (118°F) |
| Refractive Index | 1.436 |
| Quantity | 100 g |
| UN Number | UN1987 |
| Beilstein | 1719473 |
| Solubility Information | Miscible with alcohol. Immiscible with water. |
| IUPAC Name | (Z)-pent-2-en-1-ol |
| Formula Weight | 86.13 |
| Afficher plus |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.