missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Nonen-1-ol, 95%

Product Code. 11483108
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11483108

Brand: Thermo Scientific Alfa Aesar A18842.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41453-56-9 | C9H18O | 142.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41453-56-9
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00063210
InChI Key NSSALFVIQPAIQK-FPLPWBNLSA-N
Synonym cis-2-nonen-1-ol, z-2-nonen-1-ol, z-non-2-en-1-ol, unii-jlw774vc2r, 2-nonen-1-ol, z, 2z-non-2-en-1-ol, 2z-2-nonen-1-ol, jlw774vc2r, fema no. 3720, z-2-nonenol
PubChem CID 5365027
IUPAC Name (Z)-non-2-en-1-ol
SMILES CCCCCCC=CCO

Specifications

Density 0.845
Boiling Point 106°C (20 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.45
Quantity 25 g
Formula Weight 142.24
Percent Purity 95%
Chemical Name or Material cis-2-Nonen-1-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.