missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99%

Product Code. 10412251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10412251 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10412251 Supplier Thermo Scientific Acros Supplier No. 265860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5691-37-2 | C7H15NO·HCl | 165.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5691-37-2
Melting Point 131°C to 135°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C7H15NO·HCl
MDL Number MFCD00143988
Quantity 1 g
Synonym cis-2-hydroxymethyl-1-cyclohexylamine hydrochloride, cis-2-aminocyclohexyl methanol hydrochloride, 1s,2r-2-aminocyclohexyl methanol hydrochloride, +/-cis-2-hydroxymethyl-1-cyclohexylamine hydrochloride, racemic-cis-2-hydroxymethyl-1-cyclohexylamine hydrochloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
InChI Key ORPKUPNVXFLMRG-ZJLYAJKPSA-N
SMILES C1CCC(C(C1)CO)N.Cl
IUPAC Name [(1S,2R)-2-aminocyclohexyl]methanol;hydrochloride
Molecular Weight (g/mol) 165.66
PubChem CID 23545866
Formula Weight 165.66
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.