missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Hexene, 96%

Product Code. p-7035779
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15490398

Brand: Thermo Scientific Alfa Aesar H53506.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7688-21-3 | C6H12 | 84.16 g/mol

cis-2-Hexene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Hexene is used as an organic chemical synthesis intermediate.

Solubility
Immiscible in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7688-21-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00066520
InChI Key RYPKRALMXUUNKS-HYXAFXHYSA-N
Synonym cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z
PubChem CID 643835
IUPAC Name (Z)-hex-2-ene
SMILES CCC\C=C/C

Specifications

Density 0.669
Boiling Point 68°C to 70°C
Flash Point −25°C (−13°F)
Refractive Index 1.396
Quantity 5 g
UN Number UN3295
Beilstein 1719019
Solubility Information Immiscible in water.
Formula Weight 84.16
Percent Purity 96%
Chemical Name or Material cis-2-Hexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-2-Hexene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cis-2-Hexene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.