missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Hexene, 95%, Thermo Scientific™

Product Code. 10258893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10258893 1 mL Glass bottle 1mL
10483735 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10258893 Supplier Thermo Scientific Acros Supplier No. 389940010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7688-21-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00066520
InChI Key RYPKRALMXUUNKS-HYXAFXHYSA-N
Synonym cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z
PubChem CID 643835
IUPAC Name (2Z)-hex-2-ene
SMILES CCC\C=C/C

Specifications

Color Colorless
Density 0.6870g/mL
Boiling Point 68.0°C to 70.0°C
Flash Point −25°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CH(CH2)2CH3
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Quantity 1 mL
Specific Gravity 0.687
Formula Weight 84.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material cis-2-Hexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-2-Hexene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.