Learn More
cis-2-Butene-1,4-diol, 96%, remainder trans-isomer
Description
cis-2-Butene-1, 4-diol is used as a probe for studying isomerization versus hydrogenation and hydrogenolysis reactions. (DPCB-OMe) efficiently catalyzes cyclodehydration of cis-2-butene-1,4-diol with active methylene compounds to give 2-vinyl-2,3-dihydrofurans in good to high yields. Grubbs's cross metathesis of eugenol with cis-2-butene-1, 4-diol to make a natural product, an organometallic experiment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
cis-2-Butene-1, 4-diol is used as a probe for studying isomerization versus hydrogenation and hydrogenolysis reactions. (DPCB-OMe) efficiently catalyzes cyclodehydration of cis-2-butene-1,4-diol with active methylene compounds to give 2-vinyl-2,3-dihydrofurans in good to high yields. Grubbs′s cross metathesis of eugenol with cis-2-butene-1, 4-diol to make a natural product, an organometallic experiment.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 6°C to 8°C |
| Density | 1.07 |
| Boiling Point | 131°C to 132°C (12 mmHg) |
| Flash Point | 127°C (260°F) |
| Refractive Index | 1.478 |
| Quantity | 1000 g |
| Beilstein | 1679241 |
| Solubility Information | Soluble in water. |
| Formula Weight | 88.11 |
| Percent Purity | 96% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.