missing translation for 'onlineSavingsMsg'
Learn More

cis-2-(Boc-amino)cyclopropanemethanol, 97%

Product Code. 15459268
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459268 250 mg 250mg
15449268 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459268 Supplier Thermo Scientific Alfa Aesar Supplier No. H62795.MD
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 170299-61-3 | C9H17NO3 | 187.239 g/mol

cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170299-61-3
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD03844611
InChI Key OCKKMJSVLVALMF-RNFRBKRXSA-N
Synonym tert-butyl cis-2-hydroxymethyl cyclopropylcarbamate, cis-1-boc-amino-2-hydroxymethyl cyclopropane, tert-butyl cis-2-hydroxymethyl-cyclopropylcarbamate, tert-butyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, 2-methyl-2-propanyl 1r,2s-2-hydroxymethyl cyclopropyl carbamate, tert-butyl n-1r,2s-2-hydroxymethyl cyclopropyl carbamate, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, 1r, carbamic acid, 2-hydroxymethyl cyclopropyl-, 1,1-dimethylethyl ester, cis
PubChem CID 24720933
IUPAC Name tert-butyl N-[(1R,2S)-2-(hydroxymethyl)cyclopropyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CC1CO

Specifications

Quantity 250 mg
Formula Weight 187.23
Percent Purity 97%
Chemical Name or Material cis-2-(Boc-amino)cyclopropanemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.