missing translation for 'onlineSavingsMsg'
Learn More

cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%

Product Code. 15417296
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15417296

Brand: Thermo Scientific Alfa Aesar H62329.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 685508-28-5 | C13H15NO4 | 249.266 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685508-28-5
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD03844615
InChI Key ZNRUEEQIUVKKBL-MNOVXSKESA-N
Synonym cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
PubChem CID 68498468
IUPAC Name (1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
SMILES C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Quantity 250 mg
Formula Weight 249.26
Percent Purity 97%
Chemical Name or Material cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.