missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Aminocyclohexanol hydrochloride, 95%

Product Code. 10389000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10389000 1 g 1g
10722964 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10389000 Supplier Thermo Scientific Acros Supplier No. 265850010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6936-47-6 | C6H13NO·HCl | 151.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6936-47-6
MDL Number MFCD00143981
InChI Key LKKCSUHCVGCGFA-RIHPBJNCSA-N
Synonym 1r,2s-2-aminocyclohexanol hydrochloride, cis-2-aminocyclohexanol hydrochloride, 1r,2s-2-aminocyclohexan-1-ol hydrochloride, 1r,2s-rel-2-aminocyclohexanol hydrochloride, cis 1r,2s-2-amino-cyclohexanol hydrochloride, 1r,2s-2-aminocyclohexanol hcl, cyclohexanol, 2-amino-, hydrochloride, cis, cis-2-aminocyclohexanolhydrochloride, cis-2-aminocyclohexanol hcl, cis-2-amino-cyclohexanol hydrochloride
PubChem CID 12228413
IUPAC Name (1R,2S)-2-aminocyclohexan-1-ol;hydrochloride
SMILES C1CCC(C(C1)N)O.Cl

Specifications

Melting Point 186.0°C to 190.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C6H13NO·HCl
Linear Formula H2NC6H10OH·HCl
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,hot ethanol
Molecular Weight (g/mol) 151.64
Formula Weight 151.64
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material cis-2-Aminocyclohexanol hydrochloride
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.