missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%

Product Code. 11925351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
Unit Size:
10g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11925351 10 g 10g
10093450 500 mg 500mg
10607484 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11925351 Supplier Thermo Scientific Acros Supplier No. 277740100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37910-65-9 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37910-65-9
MDL Number MFCD00143982
InChI Key JWYOAMOZLZXDER-UHNVWZDZSA-N
Synonym 1r,2s-2-aminocyclopentanecarboxylic acid, cispentacin, cis-2-amino-1-cyclopentanecarboxylic acid, --cispentacin, cis-pentacin, antibiotic fr 109615, 1r,2s-2-aminocyclopentane-1-carboxylic acid, --1r,2s-cispentacin, +/--cispentacin, cispentacin,-
PubChem CID 73305
IUPAC Name (1R,2S)-2-aminocyclopentane-1-carboxylic acid
SMILES C1CC(C(C1)N)C(=O)O

Specifications

Melting Point 218.0°C to 220.0°C
Assay Percent Range 98+%
Molecular Formula C6H11NO2
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Molecular Weight (g/mol) 129.16
Quantity 10 g
Formula Weight 129.16
Percent Purity ≥98%
Chemical Name or Material cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.