missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Lupetidine, TRC

Product Code. 30482132
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482132 2.5 g 2.5g
30474855 10 g 10g
30475654 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482132 Supplier Toronto Research Chemicals Supplier No. TRCL4780102.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-17-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.2
Synonym (2R,6S)-rel-2,6-Dimethylpiperidine, cis-2,6-Dimethylpiperidine, (2R,6S)-2,6-Dimethylpiperidine, meso-2,6-Lupetidine, rel-(2R,6S)-2,6-Dimethylpiperidine
IUPAC Name (2R,6S)-2,6-dimethylpiperidine
SMILES C[C@@H]1CCC[C@H](C)N1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H15N/c1-6-4-3-5-7(2)8-6/h6-8H,3-5H2,1-2H3/t6-,7+
Formula Weight 113.1204
Chemical Name or Material cis-2,6-Lupetidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.