missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylpiperazine, 99+%

Product Code. 11332737
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11332737

Brand: Thermo Scientific Alfa Aesar L15684.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21655-48-1 | C6H14N2 | 114.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21655-48-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD07772435
InChI Key IFNWESYYDINUHV-OLQVQODUSA-N
Synonym cis-2,6-dimethylpiperazine, 2r,6s-2,6-dimethylpiperazine, cis-2,6-dimethyl piperazine, 2s,6r-2,6-dimethylpiperazine, cis-2,6-dimethyl-piperazine, piperazine, 2,6-dimethyl-, 2r,6s-rel, 2,6-dimethyl piperazine, 26dmprz, 2,6-dimethylpiperazine #, 2,6-cis-dimethylpiperazine
PubChem CID 6950261
IUPAC Name (2S,6R)-2,6-dimethylpiperazine
SMILES CC1CNCC(N1)C

Specifications

Melting Point 108°C to 111°C
Boiling Point 162°C
Flash Point 45°C (113°F)
Quantity 5 g
UN Number UN1325
Beilstein 79881
Formula Weight 114.19
Percent Purity ≥99%
Chemical Name or Material cis-2,6-Dimethylpiperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.