missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylmorpholine, Mikromol™

Product Code. 17953146
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17953146 Supplier Mikromol Supplier No. MM3511.130100
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

cis-2,6-Dimethylmorpholine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-2,6-Dimethylmorpholine
CAS 6485-55-8
Analyte or Component Names cis-2,6-Dimethylmorpholine
Shipping Condition Room Temperature
Molecular Formula C6H13NO
Certifications/Compliance ISO 9001
Synonym (2R,6S)-rel-2,6-Dimethylmorpholine, (2R,6S)-2,6-Dimethylmorpholine, (2R,6S)-2,6-Dimethylmorpholine, 2,6-cis-Dimethylmorpholine, cis-(2R,6S)-2,6-Dimethylmorpholine, cis-2,6-Dimethylmorpholine
SMILES C[C@@H]1CNC[C@H](C)O1
Molecular Weight (g/mol) 115.07
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.