missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylmorpholine, 97%

Product Code. 10630035 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10630035 100 g Glass bottle 100g
10084232 25 g Glass bottle 25g
10406963 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10630035 Supplier Thermo Scientific Acros Supplier No. 371841000
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 6485-55-8 | C6H13NO | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-55-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00078428
InChI Key HNVIQLPOGUDBSU-OLQVQODUSA-N
Synonym cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624
PubChem CID 641500
IUPAC Name (2S,6R)-2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Melting Point -85°C
Color Colorless
Density 0.9300g/mL
Boiling Point 140°C to 142°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 2% max. Trans-2,6-dimethylmorpholine (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.447
Quantity 100 g
Specific Gravity 0.93
Solubility Information Solubility in water: miscible
Viscosity 5.3 mPa.s (20°C)
Formula Weight 115.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material cis-2, 6-Dimethylmorpholine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • cis-2,6-Dimethylmorpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.