missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylmorpholine, 97%

Product Code. 11357836
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357836 50 g 50g
11347836 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357836 Supplier Thermo Scientific Alfa Aesar Supplier No. L13007.18
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6485-55-8 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-55-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00078428
InChI Key HNVIQLPOGUDBSU-OLQVQODUSA-N
Synonym cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624
PubChem CID 641500
IUPAC Name (2S,6R)-2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Melting Point -85°C
Density 0.935
Boiling Point 146°C to 147°C
Flash Point 48°C (118°F)
Refractive Index 1.446
Quantity 50 g
UN Number UN1992
Beilstein 878182
Formula Weight 115.18
Percent Purity 97%
Chemical Name or Material cis-2,6-Dimethylmorpholine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.