missing translation for 'onlineSavingsMsg'
Learn More

cis-[2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-4-yl-methyl)-1,3-dioxolan-4-yl]methyl Methanesulfonate, TRC

Product Code. 30380154
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380154 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380154 Supplier Toronto Research Chemicals Supplier No. TRCD47418310MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-[2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-4-yl-methyl)-1,3-dioxolan-4-yl]methyl Methanesulfonate
CAS 103661-14-9
Molecular Formula C14 H15 Cl2 N3 O5 S
Quantity 10 mg
Synonym Itraconazole Impurity 16
InChI Formula InChI=1S/C14H15Cl2N3O5S/c1-25(20,21)23-6-11-5-22-14(24-11,7-19-8-17-18-9-19)12-3-2-10(15)4-13(12)16/h2-4,8-9,11H,5-7H2,1H3/t11-,14+/m1/s1
SMILES CS(=O)(=O)OC[C@H]1CO[C@@](Cn2cnnc2)(O1)c3ccc(Cl)cc3Cl
IUPAC Name [(2R,4R)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-4-ylmethyl)-1,3-dioxolan-4-yl]methyl methanesulfonate
Molecular Weight (g/mol) 408.257
Formula Weight 407.011
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.