missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclopentanediol, 98%

Product Code. 10444935 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10444935 5 g Glass bottle 5g
10013750 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10444935 Supplier Thermo Scientific Acros Supplier No. 310290050
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5057-98-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5057-98-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00134192
InChI Key VCVOSERVUCJNPR-SYDPRGILSA-N
Synonym cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
PubChem CID 2733293
IUPAC Name (1R,2S)-cyclopentane-1,2-diol
SMILES C1CC(C(C1)O)O

Specifications

Melting Point 32.0°C to 33.0°C
Color Yellow
Boiling Point 83.0°C to 83.5°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C5H8(OH)2
Refractive Index 1.4770 to 1.4790
Quantity 5 g
Solubility Information Solubility in water: partially soluble in water.
Formula Weight 102.13
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material cis-1,2-Cyclopentanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.