missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanediol, 99%

Product Code. 10412855 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10412855 5 g Glass bottle 5g
10003450 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412855 Supplier Thermo Scientific Acros Supplier No. 300970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1792-81-0 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1792-81-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00064944
InChI Key PFURGBBHAOXLIO-OLQVQODUSA-N
Synonym cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol
PubChem CID 92903
ChEBI CHEBI:32329
IUPAC Name (1R,2S)-cyclohexane-1,2-diol
SMILES C1CCC(C(C1)O)O

Specifications

Melting Point 98°C to 101°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(OH)2
Quantity 5 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 116.16
Percent Purity 99%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material cis-1, 2-Cyclohexanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.