missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanedicarboxylic anhydride, 99%

Product Code. 10157703 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157703 2.5 kg Glass bottle 2.5kg
10766391 250 g Glass bottle 250g
10583701 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10157703 Supplier Thermo Scientific Acros Supplier No. 154030025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13149-00-3 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13149-00-3
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00005926
InChI Key MUTGBJKUEZFXGO-OLQVQODUSA-N
Synonym cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis
PubChem CID 641660
IUPAC Name (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES C1CCC2C(C1)C(=O)OC2=O

Specifications

Melting Point 29.0°C to 32.0°C
Color White
Density 143g/mL
Boiling Point 296.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 kg
Solubility Information Solubility in water: hydrolyzes. Other solubilities: miscible with benzene, toluene, acetone, ccl4,, chloroform, ethanol, ethyl acetate, slightly soluble in petroleum ether
Viscosity 26 mPa.s (50°C)
Formula Weight 154.17
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material cis-1, 2-Cyclohexanedicarboxylic anhydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-1,2-Cyclohexanedicarboxylic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.