missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Bis(diphenylphosphino)ethylene, 98%, ACROS Organics™

Product Code. 10297040 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
1g; Glass bottle
This item is not returnable. View return policy

Product Code. 10297040

Brand: Thermo Scientific Acros 320890010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 983-80-2
Melting Point 122°C to 126°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C26H22P2
Linear Formula (C6H5)2PCH=CHP(C6H5)2
MDL Number MFCD00003046 MFCD00063219
Quantity 1 g
Synonym cis-1,2-bis diphenylphosphino ethylene, z-1,2-bis diphenylphosphino ethene, cis-1,2-bis diphenylphosphino ethene, cis-vinylenebis diphenylphosphine, trans-1,2-bis diphenylphosphinyl ethylene, trans-vinylenebis diphenylphosphine, z-2-diphenylphosphanyl ethenyl diphenylphosphane, dppv, phosphine, trans
InChI Key NCKJIJSEWKIXAT-DQRAZIAOSA-N
SMILES C(=C\P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name [(1Z)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
Molecular Weight (g/mol) 396.41
PubChem CID 2733415
Formula Weight 396.41
Percent Purity ≥97.5%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material cis-1, 2-Bis(diphenylphosphino)ethylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.