missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,2,3,6-Tetrahydrophthalimide, 96%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| Melting Point | 131.0°C to 138.0°C |
| Color | Beige to Brown-Yellow or Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% |
| Packaging | Plastic bottle |
| Quantity | 1 kg |
| Formula Weight | 151.16 |
| Percent Purity | 96% |
| Physical Form | Powder or Flakes |
| Chemical Name or Material | cis-1,2,3,6-Tetrahydrophthalimide, 96% |
Safety and Handling
Product Identifier
- cis-1,2,3,6-Tetrahydrophthalimide
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?