missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2,3,6-Tetrahydrophthalic anhydride, 98%, ACROS Organics™

Product Code. 10347240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1kg; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10347240 1kg; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10347240 Supplier Thermo Scientific Acros Supplier No. 159720010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 935-79-5
Melting Point 98°C to 102°C
Color White
Density 156g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C8H8O3
MDL Number MFCD00005916
Quantity 1 kg
Beilstein 17, 462
Synonym cis-1,2,3,6-tetrahydrophthalic anhydride, cis-4-cyclohexene-1,2-dicarboxylic anhydride, 3ar,7as-rel-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione, unii-w9q4666nos, cis-3a,4,7,7a-tetrahydro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 1,3-isobenzofurandione, 3a,4,7,7a-tetrahydro-, cis, 3ar,7as-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione, tetrahydrophthalic anhydride, cis, 3as,7ar-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione
InChI Key KMOUUZVZFBCRAM-OLQVQODUSA-N
SMILES C1C=CCC2C1C(=O)OC2=O
IUPAC Name (3aS,7aR)-3a,4,7,7a-tetrahydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 152.149
PubChem CID 98495
Formula Weight 152.15
Percent Purity ≥97.5%
Physical Form Flakes
Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalic anhydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.