Learn More
Nickel Electro™ Clifton™ Digital Water and Oil Circulator
Product Code. 12709735
Click to view available options
Frequency:
50/60 Hz
Unit Size:
Each
Product Code. 12709735
Brand: Nickel Electro™ NE4HT
Only null left
Add to basket
Add to basket
Nickel Electro™ Clifton™ Digital Water and Oil Circulator is designed for external circulation of temperature-controlled water and heat transfer fluids in a closed circuit or maintaining a stable temperature environment in the bath for the immersion of flasks, bottles, and racks.
  • Ideal for external water circulation or immersion of flasks/bottles/racks in a highly stable temperature environment throughout the water bath.
  • Good water movement and temperature control provides temperature stability throughout the water bath.
  • Three-year warranty and comprehensive customer support.
  • Broad range of accessories available to suit wide range of applications.
  • Before leaving our factory each water bath has a unique serial number, electrically tested for safety at final inspection and a test certificate enclosed in rear of the instruction book.
  • To verify the performance of the temperature control system every unit undergoes a factory 2 point calibration procedure adjusting control to known traceable values.
  • Thermocirculator controls and painted surfaces feature anti-bacterial technology which inhibits bacterial growth, improving hygiene requirements as demanded by many clinical and laboratory environments.
  • These hygienic surfaces are part of a controlled approach to cleaner facilities.
  • Easy to clean surfaces and finishes that are resistant to routine and efficient disinfection’s.
  • The efficacy of the finish is maintained over time, as the anti-bacterial agent is integral within the finishes
Specifications
Controller Type Digital
Shipping Weight (Metric) 5 kg
Frequency 50/60 Hz