missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Circle Cover Slips, Thickness 1.5

Product Code. p-4524545 Shop All Epredia Products
Click to view available options
Diameter (Metric):
9 mm
This item is not returnable. View return policy

Product Code. 15990827

Brand: Thermo Scientific™ CB00090RAC20MNT0

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protect assay preparations from chemical attack using Thermo Scientific™ Cover Slips, Thickness 1.5, made of D 263 M colorless borosilicate glass. Made for use in medicine, biology, and research, they are available in various sizes.

Thickness 1.5:
  • 0.16 to 0.19mm
Excellent Glass Quality
  • Virtually colorless appearance
  • Very low levels of inclusions, striae, bubbles, streaks, etc
  • High spectral transmission
  • Excellent flatness
  • Finely adapted refractive index suited for microscopes
Ordering Information:
  • Packed 100 pieces in plastic boxes
  • Packing unit 1000 pieces
  • Other sizes and thicknesses available on request
TRUSTED_SUSTAINABILITY

Specifications

Thickness (Metric) #1.5 (0.16 to 0.19 mm)
Diameter (Metric) 9 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.