missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIRBP Monoclonal Antibody (6E12D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531702
This item is not returnable.
View return policy
Description
MA5-31702 has been tested in indirect ELISA.
CIRBP is cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. It acts as a translational activator and seems to play an essential role in cold-induced suppression of cell proliferation. It binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. This protein acts as a translational repressor and promotes assembly of stress granules (SGs), when overexpressed.
Specifications
| CIRBP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q14011 | |
| CIRBP | |
| Purified recombinant fragment of human CIRBP (AA: 1-90) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6E12D5 | |
| Unconjugated | |
| CIRBP | |
| A18 hnRNP; A18HNRNP; CIRBP; CIRP; cold inducible RNA binding protein; cold inducible RNA-binding protein; cold-inducible RNA-binding protein; glycine-rich RNA binding protein; glycine-rich RNA-binding protein CIRP; R74941; testicular tissue protein Li 39 | |
| Mouse | |
| Protein G | |
| RUO | |
| 1153 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction