missing translation for 'onlineSavingsMsg'
Learn More

CIQ, MedChemExpress

Product Code. 16497588
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497588 10 mg 10mg
16493606 25 mg 25mg
16440068 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497588 Supplier Medchem Express Supplier No. HY18699
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CIQ is a subunit-selective potentiator of NMDA receptors containing the NR2C or NR2D subunit.

CIQ is a subunit-selective potentiator of NMDA receptors containing the NR2C or NR2D subunit.

Specifications

Chemical Name or Material CIQ
CAS 486427-17-2
Color White
Purity Grade Notes Research
Molecular Formula C26H26ClNO5
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (106.85 mM)
SMILES O=C(C1=CC=CC(Cl)=C1)N2C(COC3=CC=C(OC)C=C3)C4=C(C=C(OC)C(OC)=C4)CC2
Molecular Weight (g/mol) 467.94
Formula Weight 467.94
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.