missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin, TRC

Product Code. 17973985
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973985 25 g 25g
17934345 50 mg 50mg
17905115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973985 Supplier Toronto Research Chemicals Supplier No. TRCC48250025G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85721-33-1
Molecular Formula C17 H18 F N3 O3
Molecular Weight (g/mol) 331.34
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-7-(1-piperazinyl)-4-oxo-3-quinoline carboxylic acid, 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, AuriPro, BAY-q 3939, Catex, Ciflafin, Cinthocip 500, Ciprine, Cipro IV, Ciprobay 100, Ciprodac, Ciprofloxacillin, Ciprofloxacin, Ciprofloxacine, Ciprolet, Ciprolet DS, Cipromed, Cipronex, Cipropol, Ciproval, Ciprovet, Ciprox, Ciproxim, Ciproxina, Cunesin, Cyclofloxacin, Euciprin, Infu-luxacin, Lipoquin, Novidate, OTO 201, Oftacifox, Otiprio, Procip, Profloxacin, Pulmaquin, Quinox XR, Urociproxin
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C17H18FN3O3/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24/h7-10,19H,1-6H2,(H,23,24)
Formula Weight 331.1332
Percent Purity >95%
Chemical Name or Material Ciprofloxacin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.