missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Piperazinyl-N4-sulfate, TRC

Product Code. 30009756
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009756 10 mg 10mg
30003730 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009756 Supplier Toronto Research Chemicals Supplier No. TRCC48252510MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105093-21-8
Molecular Formula C17 H18 F N3 O6 S
Molecular Weight (g/mol) 411.4
Synonym Sulfociprofloxacin
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-(4-sulfopiperazin-1-yl)quinoline-3-carboxylic acid
SMILES OC(=O)C1=CN(C2CC2)c3cc(N4CCN(CC4)S(=O)(=O)O)c(F)cc3C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H18FN3O6S/c18-13-7-11-14(21(10-1-2-10)9-12(16(11)22)17(23)24)8-15(13)19-3-5-20(6-4-19)28(25,26)27/h7-10H,1-6H2,(H,23,24)(H,25,26,27)
Formula Weight 411.09
Chemical Name or Material Ciprofloxacin Piperazinyl-N4-sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.