missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ciprofloxacin hydrochloride hydrate, 98%

Product Code. 15545936 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15545936 25 g 25g
15535936 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15545936 Supplier Thermo Scientific Chemicals Supplier No. 456880250
  • ONLINE EXCLUSIVE PRICE
    £76.70
    £69.65 / 25g
    Save £7.05
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ciprofloxacin hydrochloride hydrate, CAS # 128074-72-6, is the hydrate form of the hydrochloride salt of ciprofloxacin, a broad-spectrum antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ciprofloxacin HCl is a fluoroquinolone antibiotic that works by binding to bacterial DNA gyrase/DNA topoisomerase, thus inhibiting DNA replication and subsequent bacterial cell division
  • It can cause double-stranded breaks in the bacterial chromosome
  • Ciprofloxacin HCl is an analog of the antibiotic nalidixic acid
  • Works as an antibiotic against both gram-positive and gram-negative bacteria and can have fungicidal activity

Applications

  • Used for elimination of mycoplasma in cell culture
  • Can be used to study antimicrobial activity and resistance in laboratory settings
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128074-72-6
Molecular Formula C17H19ClFN3O3
Molecular Weight (g/mol) 367.81
MDL Number MFCD00242856
InChI Key DIOIOSKKIYDRIQ-UHFFFAOYSA-N
PubChem CID 62998
ChEBI CHEBI:59936
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
SMILES Cl.OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O

Specifications

Type Ciprofloxacin Hydrochloride Hydrate
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Water 4.0 to 7.0%
Quantity 25 g
Formula Weight 367.81
Percent Purity 98%
Chemical Name or Material Ciprofloxacin hydrochloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.