missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ciprofloxacin, 98%

Product Code. 13541640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13541640 25 g Glass Bottle 25g
13531640 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13541640 Supplier Thermo Scientific Chemicals Supplier No. 449620250
  • ONLINE EXCLUSIVE PRICE
    £84.10
    £75.65 / 25g
    Save £8.45
    10% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ciprofloxacin, CAS# 85721-33-1, is a second-generation fluoroquinolone showing antibiotic activity commonly used against susceptible organisms mainly gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Ciprofloxacin is considered a synthetic broad-spectrum second-generation compound with antibiotic activity mainly against gram-negative bacteria
• It inhibits bacterial DNA gyrase and topoisomerase IV, enzymes that are essential for DNA replication, cell division, and causes double-strand breaks in the bacterial chromosome

Applications

• Ciprofloxacin presents more activity against gram-negative bacteria than gram-positive bacteria.
• Ciprofloxacin does not present cross-resistance between fluoroquinolones and other antibiotics classes, which is beneficial in laboratory experiments when other antibiotics are no longer effective

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85721-33-1
Molecular Formula C17H18FN3O3
Molecular Weight (g/mol) 331.34
InChI Key MYSWGUAQZAJSOK-UHFFFAOYSA-N
Synonym ciprofloxacin, ciprofloxacine, ciprobay, ciproxan, ciprofloxacina, cipro, ciprofloxacino, ciprofloxacinum, cipro iv, bacquinor
PubChem CID 2764
ChEBI CHEBI:100241
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O

Specifications

Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 25 g
Merck Index 15,2313
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in hot dmso,soluble in hot methanol
Formula Weight 331.34
Percent Purity 98%
Chemical Name or Material Ciprofloxacin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.